Condensers, high efficient, four coil
300 mm, 45/40 NS, 45/40 NS
Catalog # LENZ05255000
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN® borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
High-efficient condensers with four cooling coils, cooling surface: 2,350 cm².
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Cone size:45/40 NS
- Jacket length:300 mm
- Socket size:45/40 NS



