Orders LinkContactUsLinkComponent
Nitrogen receivers (schlenk Tubes)
50 ml, 14/23 NS, - NS
  LENZ03370228
Nitrogen receivers (schlenk Tubes)
  LENZ03370228
Nitrogen receivers (schlenk Tubes)
Nitrogen receivers (schlenk Tubes)
50 ml, 14/23 NS, - NS
71,90 €
1 * 1 items
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With 14/23 standard ground sockets/cones and standard ground side stopcock with tube connector.
  • Cone size:
    14/23 NS
  • Socket size:
    - NS
  • Capacity:
    50 ml