Order Entry
Export
Orders LinkContactUsLinkComponent
Vydac 208TP C8, HPLC columns

Vydac 208TP C8, HPLC columns

Analytical, C8, Stainless steel, 300 Å, 5 µm, 4,60 mm, 250 mm, Silica
Catalog # 554-7581
   Low Manufacturing Impact
Supplier:   Avantor
Vydac 208TP C8, HPLC columns
Catalog # 554-7581
Supplier:  Avantor
   Low Manufacturing Impact
Vydac 208TP C8, HPLC columns
Vydac 208TP C8, HPLC columns

Analytical, C8, Stainless steel, 300 Å, 5 µm, 4,60 mm, 250 mm, Silica

Price
1 250,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.

  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications

208TP C8 is less hydrophobic than C18TP and is recommended for the analysis of peptides, 10 – 20K MW.

Vydac is a registered trademark of W.R.Grace and Co.

Specifications
  • Column Format:
    Analytical
  • Ø int.:
    4,60 mm
  • Particle Size:
    5 µm
  • Pore size:
    300 Å
  • Length:
    250 mm
  • Seq:
    19
  • Column hardware:
    Stainless steel
  • Phase:
    C8
  • Packing material:
    Silica
  • Carbon load:
    5%
  • Cat. No.:
    554-7581
  • Environmental certification:
    ISO 14001:2015 (530840 UM15)
  • Endcapped:
    Yes
  • USP code:
    L7
  • Environmentally Preferable:
    Low Manufacturing Impact