Sleeve samplers, U-D Master
Sleeve sampler set with handles, sample chambers, sampling tips and sampler case
Catalog # SAMP1500S-01
Supplier: Sampling Systems
Jump to:
Product Details & Documents
The U-D sleeve sampler was developed for accurate unit dose samples (small volume).
- Quick and easy accurate unit dose sampling
- Different volumes can be taken using the same sampler
- Simply change the tip for a different volume
Designed for use with free flowing powders and granules.
Specifications
- Description:Sleeve sampler set with handles, sample chambers, sampling tips and sampler case
- Seq:01