Orders LinkContactUsLinkComponent
Sleeve samplers, U-D Master
Sleeve samplers, U-D Master
Handle extension, 600 mm
Catalog # SAMP1500A-02
Supplier:   Sampling Systems
Sleeve samplers, U-D Master
Catalog # SAMP1500A-02
Supplier:  Sampling Systems
Sleeve samplers, U-D Master
Sleeve samplers, U-D Master
Handle extension, 600 mm
Price
152,00 €
Log in for availability
1 * 1 items
Product Details & Documents
The U-D sleeve sampler was developed for accurate unit dose samples (small volume).
  • Quick and easy accurate unit dose sampling
  • Different volumes can be taken using the same sampler
  • Simply change the tip for a different volume
Designed for use with free flowing powders and granules.
Specifications
  • Description:
    Handle extension
  • Length:
    600 mm
  • Seq:
    03