Order Entry
Export
Orders LinkContactUsLinkComponent
Sleeves for conical joints

Sleeves for conical joints

45/40
  KART1043
 :   KARTELL
Sleeves for conical joints
  KART1043
 :  KARTELL
Sleeves for conical joints
Sleeves for conical joints

45/40

 
80,50 €
Log in for availability
1 * 10 items
 

PTFE.

  • Very good chemical resistance
  • Gas, liquid and vacuum-tight (to 0,1 mbar)
  • Temperature resistant from −200 to +280 °C

For fitting onto standard ground cones. Easily detachable and suitable for repeated use. Joint grease is not needed.

Specifications
  • For NS:
    45/40