Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
BUFFER SOLUTION PH 3.788 @ 20C NIST 1 * 500 mL
BUFFER SOLUTION PH 3.788 @ 20C NIST 1 * 500 mL
Print
BUFFER SOLUTION PH 3.788 @ 20C NIST 1 * 500 mL
Catalog # REGE103788
Supplier:
REAGECON
BUFFER SOLUTION PH 3.788 @ 20C NIST 1 * 500 mL
Catalog # REGE103788
Supplier:
REAGECON
Expand image
Return to Top
BUFFER SOLUTION PH 3.788 @ 20C NIST 1 * 500 mL
Add to cart
Price
41,70 €
Log in
for availability
Unit of Measure
1 * 500 mL
-
+
Add to cart
Return Policy
Product Details & Documents