Sleeves for conical joints
12/21
Catalog # KART1037
Supplier: KARTELL
Jump to:
Product Details & Documents
PTFE.
- Very good chemical resistance
- Gas, liquid and vacuum-tight (to 0,1 mbar)
- Temperature resistant from −200 to +280 °C
For fitting onto standard ground cones. Easily detachable and suitable for repeated use. Joint grease is not needed.
Specifications
- For NS:12/21



