Order Entry
Export
Orders LinkContactUsLinkComponent
 

Sleeves for conical joints

7,5/16
  KART1035
 :   KARTELL
Sleeves for conical joints
  KART1035
 :  KARTELL
Sleeves for conical joints
Sleeves for conical joints

7,5/16

 
83,90 €
   
1 * 10 items
 

PTFE.

  • Very good chemical resistance
  • Gas, liquid and vacuum-tight (to 0,1 mbar)
  • Temperature resistant from −200 to +280 °C

For fitting onto standard ground cones. Easily detachable and suitable for repeated use. Joint grease is not needed.

 
  • For NS:
    7,5/16