Safety spectacles, Op-Tema®
Anti-fog, Blue, Grey PC
Catalog # DALL1004960
Supplier: HONEYWELL SAFETY
Jump to:
Product Details & Documents
Comfortable safety spectacles with lightweight full view lens and built-in moulded browbar.
- Soft, adjustable nose piece
- Fully adjustable
- Adjustable, break-away Flexicord retainer for added safety
Weight: 28 g
Certifications: EN 166, EN 170; 1F
Specifications
- Type:Anti-fog
- Colour:Blue
- Seq:08
- Lenses:Grey PC