Order Entry
Export
Orders LinkContactUsLinkComponent
Bellow connectors

Bellow connectors

19/38 NS, 120 mm, 19/38 NS
  COWI008.1919
 :   COWIE
Bellow connectors
  COWI008.1919
 :  COWIE
Bellow connectors
Bellow connectors

19/38 NS, 120 mm, 19/38 NS

 
65,70 €
   
1 * 1 items
 

Bellow connector, PTFE, for use as a flexible or elastic connector to prevent pressure and stresses, and to dampen vibrations in standard ground joints.

  • Inert
  • Temperature resistant up to 280 °C
  • Excellent chemical resistance

With ground joints on cone and socket. Sturdy construction with trapezoidal sealing rings, which give medium to high vacuum performance.

 
  • Cone size:
    19/38 NS
  • Socket size:
    19/38 NS
  • Total length:
    120 mm