Magnetic stirring bars, plain, PTFE
35 mm, 7 mm
Supplier: COWIE
Jump to:
Product Details & Documents
Plain PTFE magnetic stirring bars have similar performance to cylindrical bars, but provide more turbulence at low speeds.
Specifications
- Ø:7 mm
- Length:35 mm
- Seq:06



