Orders LinkContactUsLinkComponent
Connection system GL 45, DURAN®
Screw cap, GL 45, PP, with two ports GL 14
Catalog # 554-3000
Supplier:   DWK Life Sciences
Connection system GL 45, DURAN®
Catalog # 554-3000
Supplier:  DWK Life Sciences
Connection system GL 45, DURAN®
Connection system GL 45, DURAN®
Screw cap, GL 45, PP, with two ports GL 14
Price
65,60 €
Log in for availability
1 * 2 items
Product Details & Documents
Connection system with PP screw cap GL 45 with two or three PTFE ports. Flexible modular system. Typical applications: safe transfer of liquid media within a closed and sterile system (evaporation is reduced).
  • Four tubing sizes (1,6, 3,0, 3,2 or 6,0 mm) can be used by selecting the correct insert
  • Sterile pressure equalisation is possible through use of membrane filter
  • Unused ports can be provided with a blind cap
Specifications
  • Description:
    Screw cap, GL 45, PP, with two ports GL 14
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Screw cap with bore, for tube connection, GL 14, blue
    Screw cap with bore, for tube connection, GL 14, blue
    554-3004
    Log in for availability
    1 * 2 items - 9,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Insert for screw cap GL 14, 1,6 mm inner diameter
    Insert for screw cap GL 14, 1,6 mm inner diameter
    554-3005
    Log in for availability
    1 * 1 items - 15,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Screw cap, closed , with seal, GL 14, red
    Screw cap, closed , with seal, GL 14, red
    SCOT1156292
    Log in for availability
    1 * 2 items - 5,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Insert for screw cap GL 14, 3,0 mm inner diameter
    Insert for screw cap GL 14, 3,0 mm inner diameter
    554-3006
    Log in for availability
    1 * 1 items - 15,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Insert for screw cap GL 14, 3,2 mm inner diameter
    Insert for screw cap GL 14, 3,2 mm inner diameter
    554-3007
    Log in for availability
    1 * 1 items - 15,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Pressure equalization set for 2 and 3 port screw cap, incl. 0,2 μm membrane filter
    Pressure equalization set for 2 and 3 port screw cap, incl. 0,2 μm membrane filter
    554-3009
    Log in for availability
    1 * 1 items - 30,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Insert for screw cap GL 14, 6,0 mm inner diameter
    Insert for screw cap GL 14, 6,0 mm inner diameter
    554-3008
    Log in for availability
    1 * 1 items - 15,80 €