Volumetric flasks, class A, blue markings, BLAUBRAND®
5** ml, 10/19, 0,04 ± ml
612-7124
: Brand
Borosilicate glass 3.3, with PP stopper.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast blue enamel
- DE-M marking
- Individual certificate included
* In addition to DIN.
**Wide neck.
: DIN EN ISO 1042
- NS:10/19
- Capacity:5** ml
- Seq:01
- Tolerance:0,04 ± ml