Orders LinkContactUsLinkComponent
Earplugs dispenser, Leight® Source 500
Earplugs dispenser, Leight® Source 500
Earplugs without cord, refill for dispenser LS-500, 34 dB, Green
Catalog # DALL3301272
Supplier:   HONEYWELL SAFETY
Earplugs dispenser, Leight® Source 500
Catalog # DALL3301272
Supplier:  HONEYWELL SAFETY
Earplugs dispenser, Leight® Source 500
Earplugs dispenser, Leight® Source 500
Earplugs without cord, refill for dispenser LS-500, 34 dB, Green
Price
114,00 €
Log in for availability
1 * 500 PAIR
Product Details & Documents
The Leight Source 500 dispenser provides a highly visible permanent hearing protection source.
  • Heavy-duty anodized aluminum allowing constant use
  • Wall mounting for a user-friendly access
  • Crank handle to dispense earplugs
Suitable for large factories and process industries with Max®, Max Lite®, Laser Lite®, Multi Max®, Matrix™, FirmFit™, Quiet® earplugs.
Specifications
  • Colour:
    Green
  • Description:
    Earplugs without cord, refill for dispenser LS-500, 34 dB
  • Seq:
    03
  • Cat. No.:
    DALL3301272
  • Dispenser type:
    Wall-mountable
  • PPE type:
    Earplugs