Order Entry
Export
Orders LinkContactUsLinkComponent
 

Trypan blue 60% (dye content)

100 g
Catalog # A18600.22
Health
Trypan blue 60% (dye content)
Catalog # A18600.22
CAS Number:  72-57-1
Trypan blue 60% (dye content)
Trypan blue 60% (dye content)

100 g

CAS Number:  72-57-1
Price
112,00 €
Log in for availability
1 * 100 g
Product Details & Documents

CAS No.: 72-57-1 Molecular Formula: C34H24N6Na4O14S4 Formula Weight: 960.82 Melting Point: 300° Merck Reference: 14,9792 MDL No.: MFCD00003969

Used in dye exclusion studies in cell culture

Synonyms:  Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)
  • CAS Number:72-57-1
  • Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
  • MDL Number:MFCD00003969
  • Molecular Weight:960.82 g/mol
  • Merck Index:14,9792
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:200-786-7
Specifications

Specifications

  • Pk:
    100 g

Specifications