D-(+)-Maltose monohydrate 95%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
D-(+)-Maltose monohydrate 95%
Catalog # A16266.36
Supplier: Thermo Fisher Scientific
CAS Number: 6363-53-7
Jump to:
Product Details & Documents
Purity: 95% CAS No.: 6363-53-7 Molecular Formula: C12H22O11.H2O Formula Weight: 360.32 (342.30anhy) Melting Point: ca 120° dec. Solubility: Soluble in water. Slightly soluble in alcohol. Insoluble in ether Beilstein Reference: 5784659 Merck Reference: 14,5714 Optical Rotation: +130° (c=2 in water, 24h) MDL No.: MFCD00149343
Synonyms: Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate
- CAS Number:6363-53-7
- Formula:C₁₂H₂₂O₁₁·1H₂O
- MDL Number:MFCD00149343
- Molecular Weight:360.32 g/mol
- Merck Index:14,5714
- Melting Pt:∼ 120 °C (dec.)
- Storage Temperature:Ambient
- EINECS:200-716-5
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



