Order Entry
Export
Orders LinkContactUsLinkComponent
Electrolyte solutions, aqueous

Electrolyte solutions, aqueous

Electrolyte solution, L 911 storage electrolyte solution, sterilized, 1000 ml in DURAN® glass bottle, 1 L
Catalog # SCOI285138590
Supplier:   SI Analytics
Electrolyte solutions, aqueous
Catalog # SCOI285138590
Supplier:  SI Analytics
CAS Number:  
Electrolyte solutions, aqueous
Electrolyte solutions, aqueous

Electrolyte solution, L 911 storage electrolyte solution, sterilized, 1000 ml in DURAN® glass bottle, 1 L

CAS Number:  
Price
73,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Electrolyte aqueous solutions are used for reference electrodes, as electrolyte bridges and for storage.

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    Electrolyte solution, L 911 storage electrolyte solution, sterilized, 1000 ml in DURAN® glass bottle

Specifications