Drums with screw cap
10 L, 44 mm, 305 mm
Supplier: DELTALAB
Jump to:
Product Details & Documents
Drums, made of HDPE, provided with screw cap with grip. Can be used for the transport of hazardous, toxic and noxious products.
- Stackable, with security cap
- Good chemical resistance
- Four flat sides for sticking information labels
Documents
Certificates
Search by Lot NumberSpecifications
- Ø opening:44 mm
- Capacity:10 L
- Height:305 mm
- Seq:02



