Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Dimethylanisole ≥97%

2,3-Dimethylanisole ≥97%

Glass bottle for solids, 50 g
Catalog # A19992.18
Irritant
2,3-Dimethylanisole ≥97%
Catalog # A19992.18
CAS Number:  2944-49-2
2,3-Dimethylanisole ≥97%
2,3-Dimethylanisole ≥97%

Glass bottle for solids, 50 g

CAS Number:  2944-49-2
Price
185,00 €
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 97% CAS No.: 2944-49-2 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 67°(152°F) Refractive Index: 1.5200 Melting Point: 29° Boiling Point: 195° Beilstein Reference: 1859736 MDL No.: MFCD00008376

Synonyms:  1-Methoxy-2,3-dimethylbenzene
  • CAS Number:2944-49-2
  • Formula:C₉H₁₂O
  • MDL Number:MFCD00008376
  • Molecular Weight:136.19 g/mol
  • Boiling Pt:195 °C
  • Melting Pt:29 °C
  • Density:0.984
  • Flash Pt:67 °C (152 °F)
  • Storage Temperature:Ambient
  • EINECS:220-948-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications