Orders LinkContactUsLinkComponent
2,3-Dimethylanisole ≥97%
Glass bottle for solids, 10 g
  A19992.09
2,3-Dimethylanisole ≥97%
  A19992.09
 :  2944-49-2
2,3-Dimethylanisole ≥97%
2,3-Dimethylanisole ≥97%
Glass bottle for solids, 10 g
 :  2944-49-2
60,20 €
1 * 10 g
Purity: 97% CAS No.: 2944-49-2 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 67°(152°F) Refractive Index: 1.5200 Melting Point: 29° Boiling Point: 195° Beilstein Reference: 1859736 MDL No.: MFCD00008376
 :  1-Methoxy-2,3-dimethylbenzene
  •  :2944-49-2
  •  :C₉H₁₂O
  •  :MFCD00008376
  •  :136.19 g/mol
  •  :195 °C
  •  :29 °C
  •  :0.984
  •  :67 °C (152 °F)
  •  :Ambient
  •  :220-948-0
  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g