n-Heptane ≥99%
Glass bottle, 2.500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 142-82-5 Molecular Formula: CH3(CH2)5CH3 Formula Weight: 100.21 Flash Point: -4°(24°F) Refractive Index: 1.3877 Odour: Petroleum-like Melting Point: -91° Boiling Point: 97-99° Beilstein Reference: 1730763 Merck Reference: 14,4659 UN No.: 1206 MDL No.: MFCD00009544
Synonyms: high purity solvents
- CAS Number:142-82-5
- Formula:H₃C(CH₂)₅CH₃
- MDL Number:MFCD00009544
- Molecular Weight:100.20 g/mol
- Merck Index:14,4659
- Boiling Pt:97…99 °C
- Melting Pt:–91 °C
- Density:0.684
- Flash Pt:–4 °C (24 °F)
- Storage Temperature:Ambient
- EINECS:205-563-8
- UN:1206
- ADR:3,II
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2.500 mL



