Order Entry
Export
Orders LinkContactUsLinkComponent
Cyclohexanecarbonyl chloride ≥97%

Cyclohexanecarbonyl chloride ≥97%

Glass bottle for solids, 25 g
Catalog # A19824.14
Toxic
Corrosive
Cyclohexanecarbonyl chloride ≥97%
Catalog # A19824.14
CAS Number:  2719-27-9
Cyclohexanecarbonyl chloride ≥97%
Cyclohexanecarbonyl chloride ≥97%

Glass bottle for solids, 25 g

CAS Number:  2719-27-9
Price
38,20 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 97+% CAS No.: 2719-27-9 Molecular Formula: C7H11ClO Formula Weight: 146.62 Flash Point: 66°(150°F) Refractive Index: 1.4700 Boiling Point: 183-185° Beilstein Reference: 742163 UN No.: 3265 MDL No.: MFCD00001456

Synonyms:  Cyclohexanecarboxylic acid chloride, Cyclohexanecarboxylic chloride
  • CAS Number:2719-27-9
  • Formula:C₇H₁₁ClO
  • MDL Number:MFCD00001456
  • Molecular Weight:146.62 g/mol
  • Boiling Pt:183…185 °C
  • Melting Pt:29 °C
  • Density:1.088
  • Flash Pt:66 °C (150 °F)
  • Storage Temperature:Ambient
  • EINECS:220-322-7
  • UN:3265
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications