1-Tetradecanol ≥97%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 97+% CAS No.: 112-72-1 Molecular Formula: CH3(CH2)12CH2OH Formula Weight: 214.39 Flash Point: 141°(285°F) Melting Point: 37-40° Boiling Point: 289° Beilstein Reference: 1742652 Merck Reference: 14,6334 MDL No.: MFCD00004757
Synonyms: 1-Tetradecyl alcohol, 1-Hydroxy tetradecane, Myristyl alcohol
- CAS Number:112-72-1
- Formula:H₃C(CH₂)₁₂CH₂OH
- MDL Number:MFCD00004757
- Molecular Weight:214.39 g/mol
- Merck Index:14,6334
- Boiling Pt:289 °C
- Melting Pt:37…40 °C
- Density:0.823
- Flash Pt:141 °C (285 °F)
- Storage Temperature:Ambient
- EINECS:204-000-3
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



