Order Entry
Export
Orders LinkContactUsLinkComponent
1-Tetradecanol ≥97%

1-Tetradecanol ≥97%

Glass bottle for solids, 100 g
Catalog # A19638.22
Irritant
1-Tetradecanol ≥97%
Catalog # A19638.22
CAS Number:  112-72-1
1-Tetradecanol ≥97%
1-Tetradecanol ≥97%

Glass bottle for solids, 100 g

CAS Number:  112-72-1
Price
29,10 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 97+% CAS No.: 112-72-1 Molecular Formula: CH3(CH2)12CH2OH Formula Weight: 214.39 Flash Point: 141°(285°F) Melting Point: 37-40° Boiling Point: 289° Beilstein Reference: 1742652 Merck Reference: 14,6334 MDL No.: MFCD00004757

Synonyms:  1-Tetradecyl alcohol, 1-Hydroxy tetradecane, Myristyl alcohol
  • CAS Number:112-72-1
  • Formula:H₃C(CH₂)₁₂CH₂OH
  • MDL Number:MFCD00004757
  • Molecular Weight:214.39 g/mol
  • Merck Index:14,6334
  • Boiling Pt:289 °C
  • Melting Pt:37…40 °C
  • Density:0.823
  • Flash Pt:141 °C (285 °F)
  • Storage Temperature:Ambient
  • EINECS:204-000-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications