Order Entry
Export
Orders LinkContactUsLinkComponent
 

3-Methylbenzophenone ≥99%

Glass bottle for solids, 10 g
Catalog # A19568.09
3-Methylbenzophenone ≥99%
Catalog # A19568.09
CAS Number:  643-65-2
3-Methylbenzophenone ≥99%
3-Methylbenzophenone ≥99%

Glass bottle for solids, 10 g

CAS Number:  643-65-2
Price
45,20 €
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 99% CAS No.: 643-65-2 Molecular Formula: C14H12O Formula Weight: 196.25 Flash Point: 110°(230°F) Refractive Index: 1.5980 Boiling Point: 316° Beilstein Reference: 2045488 MDL No.: MFCD00008535

Synonyms:  m-Methylbenzophenone
  • CAS Number:643-65-2
  • Formula:C₁₄H₁₂O
  • MDL Number:MFCD00008535
  • Molecular Weight:196.25 g/mol
  • Boiling Pt:316 °C
  • Melting Pt:0…2 °C
  • Density:1.093
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:211-401-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications