Order Entry
Export
Orders LinkContactUsLinkComponent
Potassium dichromate ≥99%

Potassium dichromate ≥99%

Glass bottle for solids, 500 g
Catalog # A18722.36
Oxidizing
Toxic
Health
Corrosive
Environment
Potassium dichromate ≥99%
Catalog # A18722.36
CAS Number:  7778-50-9
Potassium dichromate ≥99%
Potassium dichromate ≥99%

Glass bottle for solids, 500 g

CAS Number:  7778-50-9
Price
62,20 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 7778-50-9 Molecular Formula: Cr2K2O7 Formula Weight: 294.18 Odour: Odourless Melting Point: 398° UN No.: 3087 MDL No.: MFCD00011367

  • CAS Number:7778-50-9
  • Formula:K₂Cr₂O₇
  • MDL Number:MFCD00011367
  • Molecular Weight:294.18 g/mol
  • Merck Index:14,7627
  • Boiling Pt:500 °C (1013 hPa)
  • Melting Pt:398 °C
  • Density:2.676
  • Storage Temperature:Ambient
  • EINECS:231-906-6
  • UN:3087
  • ADR:5.1,II
  • REACH:01-2119454792-32
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications