Potassium dichromate ≥99%
Glass bottle for solids, 2.500 g
Supplier: Thermo Fisher Scientific
Potassium dichromate ≥99%
Catalog # A18722.0E
Supplier: Thermo Fisher Scientific
CAS Number: 7778-50-9
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 7778-50-9 Molecular Formula: Cr2K2O7 Formula Weight: 294.18 Odour: Odourless Melting Point: 398° UN No.: 3087 MDL No.: MFCD00011367
- CAS Number:7778-50-9
- Formula:K₂Cr₂O₇
- MDL Number:MFCD00011367
- Molecular Weight:294.18 g/mol
- Merck Index:14,7627
- Boiling Pt:500 °C (1013 hPa)
- Melting Pt:398 °C
- Density:2.676
- Storage Temperature:Ambient
- EINECS:231-906-6
- UN:3087
- ADR:5.1,II
- REACH:01-2119454792-32
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:2.500 g



