Galactitol ≥97%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 608-66-2 Molecular Formula: C6H14O6 Formula Weight: 182.17 Melting Point: 187-191° Boiling Point: 275-280°/1mm Merck Reference: 14,4332 MDL No.: MFCD00064288
Synonyms: Dulcitol, rel-(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:608-66-2
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064288
- Molecular Weight:182.17 g/mol
- Merck Index:14,4332
- Boiling Pt:275…280 °C (1 mmHg)
- Melting Pt:187…191 °C
- Density:1.47
- Storage Temperature:Ambient
- EINECS:210-165-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



