Order Entry
Export
Orders LinkContactUsLinkComponent
 

3-Phenoxypropionic acid ≥98%

Glass bottle for solids, 25 g
Catalog # A17842.14
Irritant
3-Phenoxypropionic acid ≥98%
Catalog # A17842.14
CAS Number:  7170-38-9
3-Phenoxypropionic acid ≥98%
3-Phenoxypropionic acid ≥98%

Glass bottle for solids, 25 g

CAS Number:  7170-38-9
Price
171,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 7170-38-9 Molecular Formula: C6H5O(CH2)2CO2H Formula Weight: 166.17 Melting Point: 96-99° Boiling Point: 243-245° MDL No.: MFCD00002769

Synonyms:  3-Penoxypropionic acid, 3-Phenoxypropanoic acid
  • CAS Number:7170-38-9
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00002769
  • Molecular Weight:166.18 g/mol
  • Boiling Pt:243…245 °C
  • Melting Pt:96…99 °C
  • Storage Temperature:Ambient
  • EINECS:230-518-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications