2-Nitroethanol tech. 80%, Technical Grade
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
2-Nitroethanol tech. 80%, Technical Grade
Catalog # A17681.22
Supplier: Thermo Fisher Scientific
CAS Number: 625-48-9
Jump to:
Product Details & Documents
Purity: 80% CAS No.: 625-48-9 Molecular Formula: O2NCH2CH2OH Formula Weight: 91.07 Flash Point: 110°(230°F) Refractive Index: 1.4451 Melting Point: -80° Boiling Point: 194° Beilstein Reference: 1633753 MDL No.: MFCD00007405
- CAS Number:625-48-9
- Formula:C₂H₅NO₃
- MDL Number:MFCD00007405
- Molecular Weight:91.07 g/mol
- Boiling Pt:194 °C
- Melting Pt:–80 °C
- Density:1.270
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:210-895-1
Documents
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



