1,4-Benzodioxane ≥98%
Glass bottle for solids, 250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 493-09-4 Molecular Formula: C8H8O2 Formula Weight: 136.15 Flash Point: 87°(188°F) Refractive Index: 1.5490 Boiling Point: 216° Beilstein Reference: 120846 MDL No.: MFCD00006821
Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxine, 2,3-Dihydro-1,4-benzodioxin, Benzo-1,4-dioxane, 1,4-Benzodioxan
- CAS Number:493-09-4
- Formula:C₈H₈O₂
- MDL Number:MFCD00006821
- Molecular Weight:136.15 g/mol
- Boiling Pt:216 °C
- Density:1.170
- Flash Pt:87 °C (188 °F)
- Storage Temperature:Ambient
- EINECS:207-775-6
Documents
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



