Order Entry
Export
Orders LinkContactUsLinkComponent
Murexide

Murexide

Glass bottle for solids, 250 g
Catalog # A17540.30
Murexide
Catalog # A17540.30
CAS Number:  3051-09-0
Murexide
Murexide

Glass bottle for solids, 250 g

CAS Number:  3051-09-0
Price
676,00 €
Log in for availability
1 * 250 g
Product Details & Documents

CAS No.: 3051-09-0 Molecular Formula: C8H8N6O6 Formula Weight: 284.19 Melting Point: 300° Beilstein Reference: 3582175 Merck Reference: 14,6306 MDL No.: MFCD00012777

Synonyms:  Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
  • CAS Number:3051-09-0
  • Formula:C₈H₈N₆O₆
  • MDL Number:MFCD00012777
  • Molecular Weight:284.19 g/mol
  • Merck Index:14,6306
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:221-266-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications