Diethyl phthalate ≥99%
Glass bottle for solids, 1.000 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 84-66-2 Molecular Formula: C12H14O4 Formula Weight: 222.24 Flash Point: 160°(320°F) Refractive Index: 1.5020 Melting Point: -64° Boiling Point: 296-298° Beilstein Reference: 1912500 Merck Reference: 14,7371 MDL No.: MFCD00009111
Synonyms: Phthalic acid diethyl ester
- CAS Number:84-66-2
- Formula:C₁₂H₁₄O₄
- MDL Number:MFCD00009111
- Molecular Weight:222.24 g/mol
- Merck Index:14,7371
- Boiling Pt:296…298 °C
- Melting Pt:–64 °C
- Density:1.118
- Flash Pt:160 °C (320 °F)
- Storage Temperature:Ambient
- EINECS:201-550-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1.000 g



