Orders LinkContactUsLinkComponent
Potassium chlorate ≥99%
Glass bottle for solids, 250 g
  A17075.30
Potassium chlorate ≥99%
  A17075.30
 :  3811-04-9
Potassium chlorate ≥99%
Potassium chlorate ≥99%
Glass bottle for solids, 250 g
 :  3811-04-9
36,40 €
1 * 250 g
Purity: 99+% CAS No.: 3811-04-9 Molecular Formula: KClO3 Formula Weight: 122.55 Odour: Odourless Melting Point: 368° Boiling Point: 400° dec. Merck Reference: 14,7620 UN No.: 1485 MDL No.: MFCD00011361
  •  :3811-04-9
  •  :KClO₃
  •  :MFCD00011361
  •  :122.55 g/mol
  •  :14,7620
  •  :400 °C (dec.)
  •  :368 °C
  •  :2.32
  •  :Ambient
  •  :223-289-7
  •  :1485
  •  :5.1,II
  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g