Order Entry
Export
Orders LinkContactUsLinkComponent
Potassium chlorate ≥99%

Potassium chlorate ≥99%

Plastic bottle for solids, 5.000 g
Catalog # A17075.0I
Oxidizing
Irritant
Environment
Potassium chlorate ≥99%
Catalog # A17075.0I
CAS Number:  3811-04-9
Potassium chlorate ≥99%
Potassium chlorate ≥99%

Plastic bottle for solids, 5.000 g

CAS Number:  3811-04-9
Price
277,00 €
Log in for availability
1 * 5.000 g
Product Details & Documents

Purity: 99+% CAS No.: 3811-04-9 Molecular Formula: KClO3 Formula Weight: 122.55 Odour: Odourless Melting Point: 368° Boiling Point: 400° dec. Merck Reference: 14,7620 UN No.: 1485 MDL No.: MFCD00011361

  • CAS Number:3811-04-9
  • Formula:KClO₃
  • MDL Number:MFCD00011361
  • Molecular Weight:122.55 g/mol
  • Merck Index:14,7620
  • Boiling Pt:400 °C (dec.)
  • Melting Pt:368 °C
  • Density:2.32
  • Storage Temperature:Ambient
  • EINECS:223-289-7
  • UN:1485
  • ADR:5.1,II
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    5.000 g

Specifications