Order Entry
Export
Orders LinkContactUsLinkComponent
DL-α-Tocopherol (Vitamin E) ≥97%

DL-α-Tocopherol (Vitamin E) ≥97%

Glass bottle for solids, 50 g
Catalog # A17039.18
DL-α-Tocopherol (Vitamin E) ≥97%
Catalog # A17039.18
CAS Number:  10191-41-0
DL-α-Tocopherol (Vitamin E) ≥97%
DL-α-Tocopherol (Vitamin E) ≥97%

Glass bottle for solids, 50 g

CAS Number:  10191-41-0
Price
47,80 €
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 97+% CAS No.: 10191-41-0 Molecular Formula: C29H50O2 Formula Weight: 430.72 Flash Point: 240°(464°F) Physical Form: Viscous Liquid Density: 0.950 Refractive Index: 1.5050 Sensitivity: Light Sensitive Melting Point: 2-4° Boiling Point: 200-220°/0.1mm Beilstein Reference: 94012 Merck Reference: 14,10021 MDL No.: MFCD00072051

Synonyms:  (±)-α-Tocopherol, Vitamin E Alcohol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol, DL-alpha-Tocopherol, Vitamin E
  • CAS Number:10191-41-0
  • Formula:C₂₉H₅₀O₂
  • MDL Number:MFCD00072051
  • Molecular Weight:430.71 g/mol
  • Merck Index:14,10021
  • Boiling Pt:200…220 °C (0.1 mmHg)
  • Melting Pt:2…4 °C
  • Density:0.950
  • Flash Pt:240 °C (464 °F)
  • Storage Temperature:Ambient
  • EINECS:233-466-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications