Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Difluorophenol ≥98%

2,3-Difluorophenol ≥98%

Glass bottle for solids, 1 g
Catalog # A16971.03
Toxic
Corrosive
Flammable
2,3-Difluorophenol ≥98%
Catalog # A16971.03
CAS Number:  6418-38-8
2,3-Difluorophenol ≥98%
2,3-Difluorophenol ≥98%

Glass bottle for solids, 1 g

CAS Number:  6418-38-8
Price
35,20 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 98+% CAS No.: 6418-38-8 Molecular Formula: C6H4F2O Formula Weight: 130.09 Flash Point: 56°(132°F) Odour: Phenol-like Boiling Point: 53-55°/25mm Beilstein Reference: 2082265 UN No.: 2921 MDL No.: MFCD00010262

  • CAS Number:6418-38-8
  • Formula:C₆H₄F₂O
  • MDL Number:MFCD00010262
  • Molecular Weight:130.09 g/mol
  • Boiling Pt:53…55 °C (25 mmHg)
  • Melting Pt:34…38 °C
  • Flash Pt:56 °C (132 °F)
  • Storage Temperature:Ambient
  • UN:2921
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications