Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Difluorobenzaldehyde ≥97%

3,5-Difluorobenzaldehyde ≥97%

Glass bottle for solids, 5 g
Catalog # A16917.06
Flammable
Irritant
3,5-Difluorobenzaldehyde ≥97%
Catalog # A16917.06
CAS Number:  32085-88-4
3,5-Difluorobenzaldehyde ≥97%
3,5-Difluorobenzaldehyde ≥97%

Glass bottle for solids, 5 g

CAS Number:  32085-88-4
Price
98,30 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 97% CAS No.: 32085-88-4 Molecular Formula: C7H4F2O Formula Weight: 142.11 Flash Point: 56°(132°F) Refractive Index: 1.4930 Boiling Point: 61-63°/43mm Beilstein Reference: 2573392 UN No.: 1989 MDL No.: MFCD00010329

  • CAS Number:32085-88-4
  • Formula:C₇H₄F₂O
  • MDL Number:MFCD00010329
  • Molecular Weight:142.1 g/mol
  • Boiling Pt:61…63 °C (43 mmHg)
  • Flash Pt:56 °C (132 °F)
  • Storage Temperature:Ambient
  • UN:1989
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications