Order Entry
Export
Orders LinkContactUsLinkComponent
L-Threonine ≥98%

L-Threonine ≥98%

Glass bottle for solids, 25 g
Catalog # A16851.14
L-Threonine ≥98%
Catalog # A16851.14
CAS Number:  72-19-5
L-Threonine ≥98%
L-Threonine ≥98%

Glass bottle for solids, 25 g

CAS Number:  72-19-5
Price
62,20 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 72-19-5 Molecular Formula: C4H9NO3 Formula Weight: 119.12 Melting Point: 255° dec. Beilstein Reference: 1721646 Merck Reference: 14,9380 Optical Rotation: -28° (c=5 in water) MDL No.: MFCD00064270

Synonyms:  (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid
  • CAS Number:72-19-5
  • Formula:C₄H₉NO₃
  • MDL Number:MFCD00064270
  • Molecular Weight:119.12 g/mol
  • Merck Index:14,9380
  • Melting Pt:255 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-774-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications