Order Entry
Export
Orders LinkContactUsLinkComponent
 

Potassium bromide ≥99%

Glass bottle for solids, 500 g
Catalog # A16339.36
Irritant
Potassium bromide ≥99%
Catalog # A16339.36
CAS Number:  7758-02-3
Potassium bromide ≥99%
Potassium bromide ≥99%

Glass bottle for solids, 500 g

CAS Number:  7758-02-3
Price
52,10 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99+% CAS No.: 7758-02-3 Molecular Formula: KBr Formula Weight: 119.01 Refractive Index: 1.559 Odour: Odourless Boiling Point: 1435° Merck Reference: 14,7618 MDL No.: MFCD00011358

  • CAS Number:7758-02-3
  • Formula:KBr
  • MDL Number:MFCD00011358
  • Molecular Weight:119 g/mol
  • Merck Index:14,7618
  • Boiling Pt:1435 °C
  • Melting Pt:730 °C
  • Density:2.75
  • Storage Temperature:Ambient
  • EINECS:231-830-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications