Orders LinkContactUsLinkComponent
Sorbic acid ≥99%
Glass bottle for solids, 250 g
  A16196.30
Sorbic acid ≥99%
  A16196.30
 :  110-44-1
Sorbic acid ≥99%
Sorbic acid ≥99%
Glass bottle for solids, 250 g
 :  110-44-1
41,30 €
1 * 250 g
Purity: 99% CAS No.: 110-44-1 Molecular Formula: C6H8O2 Formula Weight: 112.13 Flash Point: 127°(260°F) Melting Point: 132-135° Boiling Point: 228° dec. Beilstein Reference: 1741831 Merck Reference: 14,8721 MDL No.: MFCD00002703
 :  (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid
  •  :110-44-1
  •  :CH₃CH=CHCH=CHCOOH
  •  :MFCD00002703
  •  :112.13 g/mol
  •  :14,8721
  •  :228 °C (dec.)
  •  :132…135 °C
  •  :1.205
  •  :127 °C (260 °F)
  •  :Ambient
  •  :203-768-7
  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g