Order Entry
Export
Orders LinkContactUsLinkComponent
Sorbic acid ≥99%

Sorbic acid ≥99%

Glass bottle for solids, 5.000 g
Catalog # A16196.0I
Irritant
Sorbic acid ≥99%
Catalog # A16196.0I
CAS Number:  110-44-1
Sorbic acid ≥99%
Sorbic acid ≥99%

Glass bottle for solids, 5.000 g

CAS Number:  110-44-1
Price
386,00 €
Log in for availability
1 * 5.000 g
Product Details & Documents

Purity: 99% CAS No.: 110-44-1 Molecular Formula: C6H8O2 Formula Weight: 112.13 Flash Point: 127°(260°F) Melting Point: 132-135° Boiling Point: 228° dec. Beilstein Reference: 1741831 Merck Reference: 14,8721 MDL No.: MFCD00002703

Synonyms:  (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid
  • CAS Number:110-44-1
  • Formula:CH₃CH=CHCH=CHCOOH
  • MDL Number:MFCD00002703
  • Molecular Weight:112.13 g/mol
  • Merck Index:14,8721
  • Boiling Pt:228 °C (dec.)
  • Melting Pt:132…135 °C
  • Density:1.205
  • Flash Pt:127 °C (260 °F)
  • Storage Temperature:Ambient
  • EINECS:203-768-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5.000 g

Specifications