Formaldehyde 37% (w/w) in aqueous solution stabilised
Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
Formaldehyde 37% (w/w) in aqueous solution stabilised
Catalog # A16163.AP
Supplier: Thermo Fisher Scientific
CAS Number: 50-00-0
Jump to:
Product Details & Documents
CAS No.: 50-00-0 Molecular Formula: CH2O Formula Weight: 30.03 Flash Point: 60°(140°F) Refractive Index: 1.3765 Odour: Pungent Boiling Point: 97° Beilstein Reference: 1209228 Merck Reference: 14,4235 UN No.: 1198 MDL No.: MFCD00003274
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- Merck Index:14,4235
- Boiling Pt:97 °C
- Melting Pt:–15 °C
- Density:1.083
- Flash Pt:66 °C (151 °F)
- Storage Temperature:Ambient
- UN:2209
- ADR:8,III
- :Stabilised with methanol 7-8%
- EINECS:200-001-8
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL



