Order Entry
Export
Orders LinkContactUsLinkComponent
4,7-Dichloroquinoline ≥98%

4,7-Dichloroquinoline ≥98%

Glass bottle for solids, 25 g
Catalog # A15985.14
Irritant
4,7-Dichloroquinoline ≥98%
Catalog # A15985.14
CAS Number:  86-98-6
4,7-Dichloroquinoline ≥98%
4,7-Dichloroquinoline ≥98%

Glass bottle for solids, 25 g

CAS Number:  86-98-6
Price
71,70 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 86-98-6 Molecular Formula: C9H5Cl2N Formula Weight: 198.05 Flash Point: 164°(327°F) Melting Point: 83-87° Boiling Point: 148°/10mm Beilstein Reference: 125359 MDL No.: MFCD00006774

  • CAS Number:86-98-6
  • Formula:C₉H₅Cl₂N
  • MDL Number:MFCD00006774
  • Molecular Weight:198.05 g/mol
  • Boiling Pt:148 °C (10 mmHg)
  • Melting Pt:83…87 °C
  • Flash Pt:164 °C (327 °F)
  • Storage Temperature:Ambient
  • EINECS:201-714-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications