1-Octanol ≥99%
Glass bottle, 10.000 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 111-87-5 Molecular Formula: C8H18O Formula Weight: 130.23 Flash Point: 81°(177°F) Physical Form: Liquid Density: 0.825 Refractive Index: 1.4290 Appearance: Colourless Odour: Sweet Melting Point: -16° Boiling Point: 193-195° Beilstein Reference: 1697461 Merck Reference: 14,6751 MDL No.: MFCD00002988
Synonyms: n-Octanol
- CAS Number:111-87-5
- Formula:H₃C(CH₂)₆CH₂OH
- MDL Number:MFCD00002988
- Molecular Weight:130.23 g/mol
- Merck Index:14,6751
- Boiling Pt:193…195 °C
- Melting Pt:–16 °C
- Density:0.825
- Flash Pt:81 °C (177 °F)
- Storage Temperature:Ambient
- EINECS:203-917-6
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:10.000 mL



