Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Methoxybenzonitrile ≥99%

Glass bottle for solids, 100 g
Catalog # A15945.22
Toxic
Irritant
4-Methoxybenzonitrile ≥99%
Catalog # A15945.22
CAS Number:  874-90-8
4-Methoxybenzonitrile ≥99%
4-Methoxybenzonitrile ≥99%

Glass bottle for solids, 100 g

CAS Number:  874-90-8
Price
284,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 874-90-8 Molecular Formula: C8H7NO Formula Weight: 133.15 Odour: Almond-like Melting Point: 57-61° Boiling Point: 256-257° Beilstein Reference: 508117 UN No.: 3439 MDL No.: MFCD00001818

Synonyms:  4-Cyanoanisol, Anisonitrile, p-Cyanoanisol, p-Methoxybenzonitrile
  • CAS Number:874-90-8
  • Formula:C₈H₇NO
  • MDL Number:MFCD00001818
  • Molecular Weight:133.15 g/mol
  • Boiling Pt:256…257 °C
  • Melting Pt:57…61 °C
  • Storage Temperature:Ambient
  • EINECS:212-871-6
  • UN:3439
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications