Orders LinkContactUsLinkComponent
4-Methoxybenzonitrile ≥99%
Glass bottle for solids, 25 g
  A15945.14
4-Methoxybenzonitrile ≥99%
  A15945.14
 :  874-90-8
4-Methoxybenzonitrile ≥99%
4-Methoxybenzonitrile ≥99%
Glass bottle for solids, 25 g
 :  874-90-8
115,00 €
1 * 25 g
Purity: 99% CAS No.: 874-90-8 Molecular Formula: C8H7NO Formula Weight: 133.15 Odour: Almond-like Melting Point: 57-61° Boiling Point: 256-257° Beilstein Reference: 508117 UN No.: 3439 MDL No.: MFCD00001818
 :  4-Cyanoanisol, Anisonitrile, p-Cyanoanisol, p-Methoxybenzonitrile
  •  :874-90-8
  •  :C₈H₇NO
  •  :MFCD00001818
  •  :133.15 g/mol
  •  :256…257 °C
  •  :57…61 °C
  •  :Ambient
  •  :212-871-6
  •  :3439
  •  :6.1,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g