Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,7-Octadiyne ≥98%

Glass bottle for solids, 10 g
Catalog # A15829.09
Health
Flammable
Irritant
1,7-Octadiyne ≥98%
Catalog # A15829.09
CAS Number:  871-84-1
1,7-Octadiyne ≥98%
1,7-Octadiyne ≥98%

Glass bottle for solids, 10 g

CAS Number:  871-84-1
Price
101,00 €
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 98% CAS No.: 871-84-1 Molecular Formula: HC≡C(CH2)4C≡CH Formula Weight: 106.17 Storage Temperature: 2°C to 8°C Flash Point: 23°(73°F) Refractive Index: 1.4460 Odour: Recognizable Boiling Point: 135-136° Beilstein Reference: 1697275 UN No.: 3295 MDL No.: MFCD00008580

Synonyms:  Octa-1,7-diyne
  • CAS Number:871-84-1
  • Formula:C₈H₁₀
  • MDL Number:MFCD00008580
  • Molecular Weight:106.17 g/mol
  • Boiling Pt:135…136 °C
  • Melting Pt:–32 °C
  • Density:0.810 A
  • Flash Pt:23 °C (73 °F)
  • Storage Temperature:Refrigerator
  • EINECS:212-815-0
  • UN:3295
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications