Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,5-Dihydroxybenzaldehyde ≥98%

Glass bottle for solids, 25 g
Catalog # A15565.14
Irritant
2,5-Dihydroxybenzaldehyde ≥98%
Catalog # A15565.14
CAS Number:  1194-98-5
2,5-Dihydroxybenzaldehyde ≥98%
2,5-Dihydroxybenzaldehyde ≥98%

Glass bottle for solids, 25 g

CAS Number:  1194-98-5
Price
304,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 1194-98-5 Molecular Formula: C7H6O3 Formula Weight: 138.12 Melting Point: 97-101° Beilstein Reference: 1363961

  • CAS Number:1194-98-5
  • Formula:C₇H₆O₃
  • MDL Number:MFCD00003333
  • Molecular Weight:138.12 g/mol
  • Boiling Pt:60…65 °C (12 mmHg)
  • Melting Pt:97…101 °C
  • Density:0.859 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:214-789-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications